Learn More
Medchemexpress LLC Hexanoic acid, 6-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1-oxo-1H-isoindol-4-yl]amino]- | 2338824-30-7 | 96.4% | 373.40 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY160241
Encompass_Preferred
Lenalidomide-C5-acid is a Lenalidomide-based E3 ligase ligand-linker conjugate utilized in the recruitment of CRBN protein, and it can be used to synthesize PROTACs. The product is for research use only and not sold to patients.
  • Lenalidomide-based E3 ligase ligand-linker conjugate
  • Used in the recruitment of CRBN protein
  • Can be used to synthesize PROTACs
  • Appearance: Solid
  • Color: Off-white to light yellow
Catalog No. 50-003-95173