Learn More
Medchemexpress LLC Hexanoylglycine-d11 | 24003-67-6 | 99.7% | 184.28 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY113150S10MG
Encompass_Preferred
Hexanoylglycine-d11 is the deuterium labeled Hexanoylglycine. It can be used as a tracer and as an internal standard for quantitative analysis. Deuteration, the substitution of deuterium for hydrogen, has gained attention for its potential impact on the pharmacokinetic and metabolic profiles of drugs. Stable heavy isotopes like hydrogen, carbon, and other elements are incorporated into drug molecules as tracers for quantitation during drug development.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-36214