Learn More
Medchemexpress LLC Hexanoylglycine-d2 | 1256842-52-0 | 98.0% | 175.22 | C8H13D2NO3 | 5 MG

Supplier: Medchemexpress LLC HY113150S15MG
Hexanoylglycine-d2 is the deuterium-labeled analogue of hexanoylglycine used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. Supplied as a white to off-white solid with high purity, it is intended for analytical and metabolomics workflows requiring isotopically labeled standards.
- High purity (98.0%).
- Molecular weight 175.22.
- Chemical formula C8H13D2NO3.
- Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
- Stable as powder at -20°C (3 years) or 4°C (2 years).
- Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.