Learn More
Electron Microscopy Sciences HexAuFoil is a new all-gold cryo-EM sample support technology that enables movement-free imaging to increase data quality and speed in SBDD and research.
SDP
Supplier:  Electron Microscopy Sciences Q450AR03HEX
Encompass
HexAuFoil sample supports consist of a holey gold foil on a hexagonal gold mesh grid. The grid hexagons are approximately 50 μm in size with ~10-12 μm diameter bars. The gold foil has 290 nm diameter holes arranged in a hexagonal array with 310 nm spacing, resulting in a 600 nm repeat.
Catalog No. NC3813191