Learn More
Selleck Chemical LLC Hexestrol 200mg 84-16-2 Bibenzyl
SDP
Supplier:  Selleck Chemical LLC S2473200MG
Encompass_Preferred
Hexestrol (Bibenzyl) binds to ER? and ER? with EC50 of 0.07 nM and 0.175 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-551-3