Learn More
Medchemexpress LLC Hexidium iodide 5mg | 211566-66-4 | 5 MG

Supplier: Medchemexpress LLC HY1142275MG
Hexidium iodide is a fluorescent nucleic acid stain for selectively labeling bacteria and nucleic acids in microscopy and flow cytometry. It is cell-permeant and preferentially stains gram-positive bacteria, with excitation/emission around 518/600 nm.
- Fluorescent nucleic acid stain for bacteria and cellular DNA.
- Permeant to mammalian cells and preferentially stains gram-positive bacteria.
- Excitation/emission approximately 518/600 nm for fluorescence detection.
- Soluble in DMSO; stock solutions commonly prepared at 50 mg/mL with ultrasonic assistance.
- High reported purity suitable for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.