Learn More
Cayman Chemical HexnoIc AcId-d11 1g
SDP
Supplier:  Cayman Chemical 280821 G
Encompass_Preferred
An internal standard for the quantification of hexanoic acid by GC- or LC-MS
Catalog No. 50-293-7750