Learn More
Cayman Chemical HexnoIc AcId-d11 500mg
SDP
Supplier:  Cayman Chemical 28082500 MG
Encompass_Preferred
An internal standard for the quantification of hexanoic acid by GC- or LC-MS
Catalog No. 50-293-8357