Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexokinase I Antibody (YA3853) | 102 kDa | 10UL
SDP

Supplier:  Medchemexpress LLC HYP8415610UL

Encompass_Preferred

Hexokinase I Antibody (YA3853) is a Mouse-derived, non-conjugated IgG1 monoclonal antibody targeting Hexokinase I. This antibody catalyzes the phosphorylation of various hexoses and mediates the initial step of glycolysis. It is supplied in PBS, pH 7.4.

  • Mouse-derived IgG1 monoclonal antibody
  • Targets Hexokinase I
  • Reactivity: Human, Mouse, Rat
  • Non-conjugated
  • Affinity purified
  • Molecular weight: 102 kDa
  • Applications: WB, IHC-P, ICC/IF, FC, ELISA

Catalog No. 50-004-46767


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.