Learn More
Medchemexpress LLC Hexokinase I Antibody (YA739) | affinity purified | 102 kDa | 50 UL
SDP
Supplier:  Medchemexpress LLC HYP8070250UL
Encompass_Preferred
The Hexokinase I Antibody (YA739) is a mouse-derived, non-conjugated IgG1 monoclonal antibody specifically targeting Hexokinase I. Hexokinase I phosphorylates glucose, a critical first step in glucose metabolism, and localizes to the outer membrane of mitochondria. This antibody is supplied as a liquid in PBS, pH 7.4, and is affinity purified.
  • Mouse-derived IgG1 monoclonal antibody
  • Targets Hexokinase I
  • Reacts with human, mouse, and rat
  • Non-conjugated
  • Affinity purified
  • Suitable for Western Blot (WB) applications
  • Recommended dilution ratio of 1:500-1:1000 for WB
  • Ships with blue ice
  • Store at -20°C for 1 year; avoid repeated freeze/thaw cycles
Catalog No. 50-003-95560