Learn More
Medchemexpress LLC Hexokinase II VDAC binding domain peptide, cell-permeable | 99.97% | 3997.78 | 10 MG
Supplier: Medchemexpress LLC HYP1010810MG
Hexokinase II VDAC binding domain peptide (Hxk2VBD peptide) is a cell-permeable hexokinase II VDAC binding domain. It inhibits the mitochondrial localization of hexokinase 2 (HXK2) and neurotrophic factor-directed axon outgrowth.
- Cell-permeable hexokinase II VDAC binding domain
- Inhibits mitochondrial localization of hexokinase 2 (HXK2)
- Inhibits neurotrophic factor-directed axon outgrowth
- Molecular weight of 3997.78
- Has a formula of C188H291N53O40S2
- Solid appearance, white to off-white in color