Learn More
Supply Solutions Hexylene glycol, 107-41-5, MFCD00004547, 1L
Supplier:  Supply Solutions SIGMA ARK21511L
Encompass
Linear Formula CH3CH(OH)CH2C(CH3)2OH, Molecular Weight 118.17, PharmaGrade, USP/NF, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, Synonym: (±)-2-Methyl-2,4-pentanediol, MPD
Catalog No. NC1041833