Learn More
Medchemexpress LLC Hexythiazox | 78587-05-0 | 99.6% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1851100MG
Encompass_Preferred
Hexythiazox is a selective acaricide with ovicidal, larvicidal, and nymphicidal activities. It is widely used for chemical control of mites on cotton, fruits, and vegetables. It is harmless to mammals and has no effect on beneficial insects and predators of mites.
  • Selective acaricide with ovicidal, larvicidal, and nymphicidal activities.
  • Used for chemical control of mites on cotton, fruits, and vegetables.
  • Harmless to mammals.
  • No effect on beneficial insects and predators of mites.
  • Dissipates easily in strawberries.
  • Induces toxicity in larvae of two-spotted spider mite and European red mite.
Catalog No. 50-004-10476