Learn More
Medchemexpress LLC Hexythiazox | 78587-05-0 | 99.6% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB185150MG
Encompass_Preferred
Hexythiazox is a selective acaricide that exhibits ovicidal, larvicidal, and nymphicidal activities. It is broadly utilized for the chemical control of mites affecting cotton, fruits, and vegetables, and is considered harmless to mammals while having no adverse effects on beneficial insects or mite predators.
  • Selective acaricide activity
  • Ovicidal, larvicidal, and nymphicidal actions
  • Effective for mite control on cotton, fruits, and vegetables
  • Harmless to mammals
  • No adverse effect on beneficial insects or predators
Catalog No. 50-004-10912