Learn More
ATCC HEY-T30; Ovarian Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL3252
Encompass
HEY-T30 is a Taxol resistant derivative of an epithelial-like cell line that was isolated in 2006. The parental cell line, HEY, was derived from a human ovarian cancer xenograft originally grown from a peritoneal sample from a patient with moderately differentiated papillary cystadenocarcinoma of the ovary. CRL-3252, HEY-T30, is a derivative of the HEY. It was developed by exposure of its parental cell line, HEY, to stepwise escalating concentrations of Taxol over a 6-month period and it is maintained in medium containing 30 nmol/L Taxol. This cell line was deposited by Jurriaan Brouwer-Visser Institution and can be used in cancer research.
Catalog No. 50-238-2996