Learn More
Apexbio Technology LLC HyperFluor 594 TSA Fluorescence System Kit 200slides
SDP
Supplier:  Apexbio Technology LLC K4302200
Encompass_Preferred
Our Tyramide Signal Amplification (TSA) system can detect low-abundance targets in fluorescent immunocytochemistry (ICC) immunohistochemistry (IHC) and in situ hybridization (ISH) techniques enhancing signal sensitivity up to 100-fold The TSA fluorescence kits utilize horseradish peroxidase (HRP) to directly catalyze the covalent deposition of fluorescent groups near the immobilized enzyme The labeling process is rapid (under 10 min) and can be directly visualized through standard or confocal microscope TSA fluorescence kits can be used in combination with anti-fluorescence protocols TSA technology is suitable for brightfield microscopy applications with enzyme conjugates and appropriate chromogenic substrates This kit provides fluorescence labeling with HyperFluor 594 (Ex/Em 591 nm/617 nm) which can be easily detected using standard red/Texas Red filter sets
Catalog No. 50-310-7053