Learn More
Perkin Elmer US LLC HF-Resistant Sample Introduction Kit with Sapphire Injector for NexION 300/350
Supplier:  Perkin Elmer US LLC N8142002
Encompass_Preferred
Complete HF-resistant sample introduction system for NexION 300 and 350. All wetted surfaces are PTFE and Sapphire and it provides a chemically inert and clean introduction system. This kit includes: 47 mm PFA Barrel Spray Chamber with Endcap (N8142000) PFA-ST Nebulizer (N8122192) 1.8 mm I.D. Sapphire Injector (N8122411) 0-Slot Quartz Torch (N8122006) 0.5 mm I.D. Nebulizer Capillary Tubing (N8122384) 0.25 mm I.D. Nebulizer Capillary Tubing (N8122383) Spray Chamber Threaded Drain Line (N8141425)
Catalog No. 50-210-4770
Only null left
Add to Cart
Request a Quote