Learn More
Medchemexpress LLC HG-10-102-01 | 1351758-81-0 | MFCD28160565 | 99.6% | 377.83 g/mol | C17H20ClN5O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY134885MG
Encompass_Preferred
HG-10-102-01 is a potent, selective, brain-penetrant small-molecule inhibitor of leucine-rich repeat kinase 2 (LRRK2) for research use. It exhibits low-nanomolar activity against wild-type and G2019S mutant LRRK2 and is supplied as a high-purity solid suitable for cellular and in vivo studies.
  • Potent LRRK2 inhibition at low nanomolar concentrations.
  • Brain-penetrant profile suitable for central nervous system studies.
  • Active against both wild-type and G2019S mutant forms of LRRK2.
  • High chemical purity appropriate for research applications.
  • Supplied in milligram-scale quantities for laboratory use.
  • Molecular formula C17H20ClN5O3; molecular weight 377.83 g/mol.
Catalog No. 50-002-22726