Learn More
Selleck Chemical LLC HG-9-91-01
SDP
Supplier:  Selleck Chemical LLC S839310MM/1ML
Encompass_Preferred
HG-9-91-01 (SIK inhibitor 1) is a potent and highly selective inhibitor of salt-inducible kinase (SIK) with IC50 of 0 92 nM 6 6 nM and 9 6 nM for SIK1 SIK2 and SIK3 respectively
Catalog No. 50-000-01561