Learn More
TARGETMOL CHEMICALS INC HG-9-91-01 5mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T45995MG
Encompass_Preferred
HG-9-91-01 is a potent and highly selective salt-inducible kinase (SIKs) inhibitor with IC50s of 0.92 nM, 6.6 nM and 9.6 nM for SIK1, SIK2 and SIK3respectively.
Catalog No. NC1624939