Learn More
TARGETMOL CHEMICALS INC HG-9-91-01 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4599
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HG-9-91-01 (SIK inhibitor 1) is a potent and highly selective salt-inducible kinase (SIKs) inhibitor with IC50s of 0.92 nM 6.6 nM and 9.6 nM for SIK1 SIK2 and SIK3respectively. purity: 99%
Catalog No. 50-257-9865