Learn More
Selleck Chemical LLC HG106-E1080-25MG
SDP
Supplier:  Selleck Chemical LLC E108025MG
Encompass_Preferred
HG106 a potent SLC7A11 inhibitor leads to enhanced ROS generation ER stress and ultimately growth arrest specifically in KRAS-mutant lung adenocarcinoma (LUAD)
Catalog No. 50-313-7871