Learn More
Medchemexpress LLC HGFA/HGF Activator 10ug | 10ug
SDP
Supplier:  Medchemexpress LLC HYP7796010UG
Encompass_Preferred
HGFA/HGF activating proteins activate HGF by converting HGF into its heterodimeric form which is critical for cell growth development and tissue repair It induces cellular responses such as proliferation migration and differentiation HGFA/HGF Activator Protein Mouse (HEK293 His) is the recombinant mouse-derived HGFA/HGF Activator protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-95142