Learn More
Cayman Chemical HGlyArgpNAhydrochLRIde 10mg
SDP
Supplier:  Cayman Chemical 2770510 MG
Encompass_Preferred
A colorimetric thrombin substrate; cleavage of the GR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of thrombin activity
Catalog No. 50-293-9965