Learn More
Medchemexpress LLC HHS-0701 | 2851993-91-2 | 98.9% | 396.46 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13866550MG
Encompass_Preferred
HHS-0701 is a sulfur-triazole exchange (SuTEx) ligand and a potent tyrosine-reactive prostaglandin reductase 2 (PTGR2) inhibitor. It works by blocking PTGR2 metabolism of the lipid substrate 15-Keto-PGE2.
  • Potent PTGR2 inhibitor
  • Blocks PTGR2 metabolism of 15-Keto-PGE2
  • Shows concentration-dependent blockade of probe labeling in recombinant PTGR2 overexpressing HEK293T cells
Catalog No. 50-003-47885