Learn More
TARGETMOL CHEMICALS INC HHS-0701 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T677665MG
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HHS-0701 is a sulfur-triazole exchange (SuTEx) ligand. HHS-0701 is a potent inhibitor of tyrosine-reactive prostaglandin reductase 2 (PTGR2). HHS-0701 can block PTGR2 metabolism of the lipid substrate 15-Keto-PGE2. purity: 97%
Catalog No. 50-257-9258