Learn More
Electron Microscopy Sciences Hi/Lo Cloth Temperature Tape, 1" Wide 40 YD
SDP
Supplier:  Electron Microscopy Sciences 7701210
Encompass_Preferred
These tapes mold tightly to any container, withstands acids, alkalis, oils and greases. It's will not crack, peel, or fall off in 550 degree F dry oven temperatures. Hi/Lo Temperature tape also sticks tightly. This tape must be cut with scissors.
Catalog No. 50-312-94