Learn More
Electron Microscopy Sciences Hi/Lo Write-On™ Temperature Tape 3/4", White, 1" Core 15 YD
SDP
Supplier:  Electron Microscopy Sciences 77009W2
Encompass_Preferred
These color-coded label tapes work well with all glass and plastic labware, racks, tubing, bags, boxes, and cabinets. It adheres to any surface and peels off without leaving a sticky residue. Tapes resist moisture and most solvents and can withstand autoclaving (250 degree F). It may be used as well down to -73 degree C.
Width: 3/4"
Color: White
Core: 1"
Catalog No. 50-299-34