Learn More
Medchemexpress LLC HY-101550 5mg Medchemexpress, HI-TOPK-032 CAS:487020-03-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1015505MG
Encompass_Preferred
Medchemexpress, HY-101550 5mg HI-TOPK-032 CAS:487020-03-1 HI-TOPK-032 is a potent and specific TOPK inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2164