Learn More
Cole-Parmer AEMC 275HVD Hi-Voltage Detector Noncontact 240v/275kv
Supplier:  Cole-Parmer 2001983
Encompass
Non-contact with high voltage detector
FEATURES
  • Non-contact with high voltage
  • Wide range of detection; finds 80V to 275kV; one instrument fits all applications
  • Eight switch selectable voltage ranges (AE213112)
  • Ten switch-selectable voltage sensitivity ranges (AE213113)
  • Lightweight, minimizes sway at the end of long hot sticks
  • Self-test position ensures that all system functions and indicators are working properly by energizing the complete system
  • Audible and visual indication of live voltage
  • Automatic self-test beeps every two seconds (AE213113)
  • Digital filter rejects signals outside of 40 to 70 Hz
  • Designed for use with hot sicks incorporating a universal spline adapter
Catalog No. 50-992-627