Learn More
Mac-Mod Avantor Hichrom RPB, 110 A, 5 um, 4.6 x 250 mm HPLC Column
SDP
Supplier:  Mac-Mod HIRPB250A
Encompass
These columns offer high performance in order to tackle the most challenging reversed-phase applications. The use of ultra-pure silica, advanced bonding technology, superior column specifications and comprehensive batch validation all contribute to the columns excellent reproducibility. They are suitable for acidic, basic and neutral molecules and available in 3.5 and 5 ?m particle sizes. Guard cartridges are available for column dimensions, to further extend column lifetime by preventing irreversible adsorption and frit blockage at the top of the column. Note: Use with freestanding holder HI-161 and column coupler HI-081.
Catalog No. 50-900-02148