Learn More
Medchemexpress LLC HIF-1/2α-IN-2 | 862974-22-9 | 99.6% | 342.35 g·mol⁻¹ | C16H11FN4O2S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15134410MG
Encompass_Preferred
HIF-1/2α-IN-2 is a small-molecule inhibitor of hypoxia-inducible factors HIF-1α and HIF-2α that reduces cellular HIF levels by targeting the iron-sulfur cluster assembly protein ISCA2, producing an iron-starvation response. It is used in research to study HIF-dependent signaling, iron-sulfur cluster biology, and ferroptosis-related mechanisms.
  • Inhibits HIF-1α and HIF-2α by targeting ISCA2.
  • Induces an iron starvation response in cells.
  • Useful for studying HIF signaling and iron-sulfur cluster biology.
  • Supports research into ferroptosis and related cell-death pathways.
  • Available in small research quantities suitable for in vitro studies.
Catalog No. 50-002-87549