Learn More
Medchemexpress LLC HIF-2α agonist 2 | 2750141-15-0 | C13H8Br2N2O2S | 10MM/1ML
SDP
Supplier:  Medchemexpress LLC HY15301610MM/1ML
Encompass_Preferred
HIF-2α agonist 2 is a chemical compound targeting HIF/HIF Prolyl-Hydroxylase. It is non-cytotoxic against 786-O-HRE-Luc cells and can be used for oxygen metabolism research. This compound is supplied as a solid.
  • Targets HIF/HIF Prolyl-Hydroxylase
  • Suitable for oxygen metabolism research
  • Non-cytotoxic against 786-O-HRE-Luc cells
  • Has an EC50 value of 1.68 μM at the dose of 20 μM
  • Supplied as a solid
Catalog No. 50-003-72515