Learn More
Medchemexpress LLC HIF-2α agonist 2 | 2750141-15-0 | C13H8Br2N2O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1530165MG
Encompass_Preferred
HIF-2α agonist 2 is an agonist that activates HIF-2 with an EC50 value of 1.68 μM at a dose of 20 μM. It is non-cytotoxic against 786-O-HRE-Luc cells and can be used for oxygen metabolism research.
  • Activates HIF-2 with an EC50 value of 1.68 μM
  • Non-cytotoxic against 786-O-HRE-Luc cells
  • Useful for oxygen metabolism research
Catalog No. 50-003-73765