Learn More
Meso Scale Discovery HIF1alpha Whole Cell Lysate
SDP
Supplier:  Meso Scale Discovery K150DKD1
Encompass
The HIF-1alpha Whole Cell Lysate Kit from Mesoscale Discovery is used to detect HIF-1alpha protein in cell lysates The kit contains a 96-well plate coated with capture antibody detection antibody sample diluent wash buffer readout buffer and a vial of lyophilized recombinant HIF-1alpha standard The assay is performed by adding sample or standard detection antibody washing adding readout buffer and analyzing on an electrochemiluminescence plate reader The kit can quantitatively measure HIF-1alpha levels from cell lysates with high sensitivity and wide dynamic range
Catalog No. 50-000-81850