Learn More
Selleck Chemical LLC Higenamine hydrochloride S3960-5mg
SDP
Supplier:  Selleck Chemical LLC S39605MG
Encompass_Preferred
Higenamine (Norcoclaurine ( -)-Demethylcoclaurine) also known as Norcoclaurine HCl is a non-selective 2 adrenoceptor agonist which is a chemical compound naturally occurring in a number of plants
Catalog No. 50-310-9588