Learn More
Genscript Corporation High Affinity Ni-Charged Resin
Supplier:  Genscript Corporation L00223500
Encompass
GenScript High Affinity Ni-Charged Resin is an 4% cross-linked agarose medium covalently coupled to a chelating agent that binds Ni2 by four coordination sites for high-affinity purification of polyhistidine-tagged recombinant proteins High Affinity Ni-Charged Resin has low Ni2 leakage high protein-binding capacity and stability and is compatible with a wide range of additives used in protein purification This makes High Affinity Ni-Charged Resin the excellent choice for high performance purification of polyhistidine-tagged proteins
Catalog No. 50-000-58389