Learn More
XenoTech,LLC High AO/XO Pooled Human Liver Cytosol: Mixed GenderSpecies: HumanGender: MixedProtein Concentration: 10 mg/mLPool Size: 20Unit Of Measure: 1mL
SDP
Supplier:  XenoTech,LLC H0606.C(AX)
Encompass_Preferred
High AO/XO Pooled Human Liver Cytosol: Mixed GenderSpecies: HumanGender: MixedProtein Concentration: 10 mg/mLPool Size: 20Unit Of Measure: 1mL
Catalog No. NC1779772