Learn More
Alkali Scientific High-Efficiency Erlenmeyer Shake Flask, PC, Flat Base, 3L, Wide-Mouth, 4/Case, Vented Cap
SDP
Supplier:  Alkali Scientific EFN608W
Encompass_Preferred
High Efficiency Erlenmeyer Flask With ISO9001&13485 certified, High Efficiency Erlenmeyer Flask are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.  Material: PC Bottle & HDPE Cap & Nylon Handle, Vent caps with 0.2um hydrophobic filters for gas exchange without contamination.   Product Feature: • There’s a scale made by injection molding on the flask body to facilitate observing the filled liquid volume.• Sterilized by E-beam, SAL=10-6.• Non-Pyrogenic, DNase/Rnase free.• Individually packaged in sterile bag.   Application: Ideal for shaker culture applications.   Tips:· Recommended volume :30-40% of the bottle volume;· Rotation speed suggestion: the starting speed is 75-125RPM, you can control the rotation speed according to the situation;· The liquid shake shaker should pay attention to the water level, the gas shake shaker should pay attention to the temperature
Catalog No. 50-302-7488