Learn More
KRISHGEN BIOSYSTEMS Enzyme Immunoassay for the Quantitative Determination of H5 Host Cell Proteins in cell culture supernatant and biological solutions. Assay Range: 3 – 100 ng/ml; Sensitivity: 3 ng/ml
Supplier:  KRISHGEN BIOSYSTEMS KBBP21
Encompass_Preferred
Pack Size: 1 x 96 wells.Introduction:High Five cells (sometimes referred to as High-Five or Hi5) represent a safe, effective and inexpensive platform for protein production. Indeed, their remarkable ability to produce very large amounts of recombinant proteins – such as diagnostic reagents and recombinant vaccines – remains unmatched. The baculovirus expression vector system (BEVS) in insect cells offers highly scalable and inexpensive protein production. High Five cells and sub-clones have been reported to produce 2-10x high levels of recombinant proteins compared to Sf9/Sf21 insect cells. BTI is the sole proprietor of High Five cells and related sub-clones.The KRIBIOLISA H5 (High Five) HCP ELISA kit is designed to quantitatively measure HCPs contamination in pharmaceutical products manufactured using the High Five Cells expression systems. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Lot to Lot Consistency with Precision CV <15%.
Catalog No. NC2747155