Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P High-Performance Pump Head for High-Performance Precision Tubing, Polyester and SS Housing, SS Rotor
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX7760062/DEL
Encompass
Featuring the highest flow rates, best pressure and suction performance of any I/P pump head. Polyester and stainless steel (SS) construction withstands tough process environments. Versatile mounting configurationsmount with tubing exiting to the top or side for flexible setup. C-shaped design allows tubing to enter and exit the same side of the pump head; facilitates connection to tubing inlet and outlet. Adjustable tubing retention; adjust to prevent tubing movement in head.
Catalog No. NC9578261