Learn More
LIEBHERR USA High‑Performance Refrigerator available in 50-cubic-foot
Supplier:  LIEBHERR USA SRPVH140213720
Encompass
Glass/steel door high-volume refrigerators with forced air cooling. White steel exterior and stainless-steel interior. Choose between two temperature stability options to suit differing laboratory requirements. The intuitive user interfaces provide useful extended multi-functionality, e.g. a maintenance timer, and the LED interior lighting ensures good visibility of the contents of these large appliances. Advanced connectivity options to suit varying requirements for simplified data storage, temperature control and alarm signals. These advanced lab fridges also feature a +2 °C safety device to protect sensitive contents from freezing. Perfection series ±2 K deviation from setpoint and DIN 13277 compliance. Both with calibration function.A ll models are available in Perfection e series. Glass door versions are optimised for improved display area and a better overview of the contents. Wire shelves with 60 kg shelf load
Catalog No. NC3977282