Learn More
Harrick Plasma High Power Expanded Plasma Cleaner 115V
SDP
Catalog No. NC0696964
Encompass_Preferred
The PDC-001-HP High Power Expanded Plasma Cleaner is a versatile instrument, suitable for etching organic thin films (10-100 nm thickness) as well as nanoscale cleaning and surface modification. Maximum RF power of 45W; Adjustable RF power settings (LOW, MEDIUM, HIGH); LOW RF power is equivalent to HIGH RF power setting on PDC-001; 6in. diameter x 6.5in. length Pyrex chamber; Hinged door with viewing window; Active fan cooling; Integral switch for a vacuum pump; Metering valve to qualitatively control gas flow and vacuum pressure; Three-way valve to quickly switch from bleeding in gas, isolating the chamber, and venting; Size: 11in. H x 18in. W x 9in. D; Weight: 37 lb. REQUIRED: A separate vacuum pump (minimum pump speed 23 L/min, ultimate total pressure of 200 mTorr or less) is required to run the plasma process.
Catalog No. NC0696964 Supplier Harrick Plasma Supplier No. PDC001HP
Add to Cart
Add to Cart
Request a Quote
The PDC-001-HP High Power Expanded Plasma Cleaner is a versatile instrument, suitable for etching organic thin films (10-100 nm thickness) as well as nanoscale cleaning and surface modification. Maximum RF power of 45W; Adjustable RF power settings (LOW, MEDIUM, HIGH); LOW RF power is equivalent to HIGH RF power setting on PDC-001; 6in. diameter x 6.5in. length Pyrex chamber; Hinged door with viewing window; Active fan cooling; Integral switch for a vacuum pump; Metering valve to qualitatively control gas flow and vacuum pressure; Three-way valve to quickly switch from bleeding in gas, isolating the chamber, and venting; Size: 11in. H x 18in. W x 9in. D; Weight: 37 lb. REQUIRED: A separate vacuum pump (minimum pump speed 23 L/min, ultimate total pressure of 200 mTorr or less) is required to run the plasma process.