Learn More
Pendotech Pendotech Pressuremat Monitor, Transmitter, And Alarm (Pmat-Shr)
Supplier:  Pendotech PMATSHR
Encompass
High Resolution PressureMAT monitor, transmitter, alarm for 1 single use pressure sensor (analog 4-20 mA output, alarm relay output, tare input)
Catalog No. NC1831846