Learn More
Nanocomposix BioReady High Sensitivity Gold Conjugation Kit for Lateral Flow - 150 nm Gold Nanoshells
SDP
Supplier:  Nanocomposix GSZR15010M
Encompass_Preferred
Imagine an optimized system with time-after-time consistency that provides increased sensitivity and saves you time and money!The High Sensitivity Conjugation Kit is a complete “classroom-in-a-box” with detailed protocols to help you produce robust covalently-bound conjugates. This kit is available with our ultra-bright 150 nm gold nanoshells as the reporter particle and includes everything you need to optimize your conjugations.Benefits of the kit include:-Pre-validated consumables eliminate the risk of purchasing non-compatible reagents, saving weeks to months of unnecessary troubleshooting-Increase your assay sensitivity up to 20-fold with 150 nm gold nanoshells compared to 40 nm gold* nanoparticles-Reduces antibody costs up to 65% compared to physisorption-Saves time by reducing the number of conjugations necessary to find optimal antibody pairs
Catalog No. NC1820746