Learn More
Agilent Technologies High Sensitivity RNA ScreenTape
Supplier:  Agilent Technologies 50675579
Encompass
High Sensitivity RNA ScreenTape. For the high sensitivity analysis of total RNA down to a sensitivity of 100 pg/µL. Includes 7 ScreenTape devices.
Catalog No. NC1617771