Learn More
IBI SCIENTIFIC 3P Hi-Speed Mini Plasmid Kit, 300 preps
Supplier:  IBI SCIENTIFIC 3P IB47102
Encompass_Preferred
High-Speed Plasmid Mini Kit - 300 preps Sample: 1-4ml of bacterial culture Format: Spin column Operation: Centrifuge or vaccum manifold Binding Capacity: Up to 30ug Expected Yield: 20-30ug for high-copy plasmid 3-10ug for low-copy plasmid.
Catalog No. NC0645830