Learn More
Nelco Products Inc HIGH-TEMP CABLE TIE, NYLON 4/6
Supplier:  Nelco Products Inc N850PA469CQTY 1024
Encompass_Preferred
8” long, 50lb tensile (.190”wide), High-temp Cable Tie, Nylon 4/6, Natural, 100/pkg Temp rating: -40F to 275F
Catalog No. NC1986840