Learn More
Chemglass Life Sciences High Temperature Cap, GL-32, Solid, PBTE
SDP
Supplier:  Chemglass Life Sciences CLS148601
Encompass_Preferred
High temperature PBTE cap will withstand temperature from -45 to 200 degreesC. complete with PTFE seal.
Catalog No. 50-273-3288