Learn More
Lakeland Industries High Visibility
Supplier:  Lakeland Industries V8AM0312ZL3X
Encompass_Preferred
Lakeland Class 2 FR Modacrylic Mesh Hi-Vis Vests are designed with inherently FR mesh fabric, and reflective material. V8AM0322VL meets ANSI 107-2015, Class 2, with open sides, adjustable side snaps, and zipper closure. Available in sizes MD, XL, 3X, 5X, and 7X, these vests are ideal for worker environments where visibility is important, and when flash fire is a concern.
Catalog No. 50-284-1145